Assay Details
Gene Family ID: | MIPF0000209, mir-362 |
Mature miRNA Sequence: | AACACACCUGUUCAAGGAUUCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-362-3p | MIMAT0004684 |
| Rat | rno-miR-362-3p | MIMAT0017357 |
Stem-loop Details
Mouse
Rat

Genomic Map