Assay Details
Amplicon Length: | 89 |
Chromosome Location: | Chr. 12 - 109617956 - 109617956 [-] on 109617956 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-370-5p | MIMAT0017174 |
Mouse | mmu-miR-370-3p | MIMAT0001095 |
Stem-loop Details
Mouse
