Assay Details
Amplicon Length: | 62 |
Chromosome Location: | Chr. 12 - 109594113 - 109594113 [-] on 109594113 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-434-5p | MIMAT0001421 |
Mouse | mmu-miR-434-3p | MIMAT0001422 |
Stem-loop Details
Mouse
