Assay Details
Gene Family ID: | MIPF0000439, mir-434 |
Mature miRNA Sequence: | UUUGAACCAUCACUCGACUCCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-434-3p | MIMAT0001422 |
Rat | rno-miR-434-3p | MIMAT0005315 |
Stem-loop Details
Mouse
Rat
