Assay Details
Amplicon Length: | 72 |
Chromosome Location: | Chr. 13 - 113035789 - 113035789 [-] on 113035789 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-449c-5p | MIMAT0003460 |
Mouse | mmu-miR-449c-3p | MIMAT0022715 |
Stem-loop Details
Mouse
