Assay Details
Gene Family ID: | MIPF0000133, mir-449 |
Mature miRNA Sequence: | AGGCAGUGCAUUGCUAGCUGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-449c-5p | MIMAT0003460 |
Rat | rno-miR-449c-5p | MIMAT0017803 |
Stem-loop Details
Mouse
Rat
