Assay Details
Amplicon Length: | 113 |
Chromosome Location: | Chr. X - 16117022 - 16117022 [-] on 16117022 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-362-3p | MIMAT0017357 |
| Rat | rno-miR-362-5p | MIMAT0012828 |
Stem-loop Details
Rat

Genomic Map