Assay Details
Amplicon Length: | 91 |
Chromosome Location: | Chr. 2 - 44895922 - 44895922 [+] on 44895922 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-449c-5p | MIMAT0017803 |
Rat | rno-miR-449c-3p | MIMAT0017804 |
Stem-loop Details
Rat
