Assay Details
Gene Family ID: | MIPF0000144, mir-317 |
Mature miRNA Sequence: | UGAACACAGCUGGUGGUAUCCAGU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Drosophila melanogaster | dme-miR-317-3p | MIMAT0000409 | |
Drosophila ananassae | dan-miR-317 | MIMAT0008452 | |
Drosophila erecta | der-miR-317 | MIMAT0008541 | |
Drosophila grimshawi | dgr-miR-317 | MIMAT0008586 | |
Drosophila sechellia | dse-miR-317 | MIMAT0008789 | |
Drosophila simulans | dsi-miR-317 | MIMAT0008863 | |
View More Rows | |||
Drosophila virilis | dvi-miR-317-3p | MIMAT0008947 | |
Drosophila willistoni | dwi-miR-317 | MIMAT0009024 | |
Drosophila yakuba | dya-miR-317 | MIMAT0009055 |
Stem-loop Details
Drosophila melanogaster
Drosophila ananassae
Drosophila erecta
Drosophila grimshawi
Drosophila sechellia
Drosophila simulans
Drosophila virilis
Drosophila willistoni
Drosophila yakuba

More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-317 (v16)
Drosophila virilis: dvi-miR-317 (v20)
Drosophila virilis: dvi-miR-317 (v20)
