Assay Details
Gene Family ID: | MIPF0000144, mir-317 |
Mature miRNA Sequence: | UGAACACAGCUGGUGGUAUCCAGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-317-3p | MIMAT0000409 |
| Drosophila ananassae | dan-miR-317 | MIMAT0008452 |
| Drosophila erecta | der-miR-317 | MIMAT0008541 |
| Drosophila grimshawi | dgr-miR-317 | MIMAT0008586 |
| Drosophila sechellia | dse-miR-317 | MIMAT0008789 |
| Drosophila simulans | dsi-miR-317 | MIMAT0008863 |
| View More Rows | ||
| Drosophila virilis | dvi-miR-317-3p | MIMAT0008947 |
| Drosophila willistoni | dwi-miR-317 | MIMAT0009024 |
| Drosophila yakuba | dya-miR-317 | MIMAT0009055 |
Stem-loop Details
Drosophila melanogaster
Drosophila ananassae
Drosophila erecta
Drosophila grimshawi
Drosophila sechellia
Drosophila simulans
Drosophila virilis
Drosophila willistoni
Drosophila yakuba
More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-317 (v16)
Drosophila virilis: dvi-miR-317 (v20)
Drosophila virilis: dvi-miR-317 (v20)

Genomic Map