Assay Details
Gene Family ID: | MIPF0000015, MIR399 |
Mature miRNA Sequence: | UGCCAAAGGAGAUUUGCCCUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Arabidopsis | ath-miR399a | MIMAT0000951 | |
| Arabidopsis lyrata | aly-miR399f-3p | MIMAT0017582 | |
| Cynara cardunculus | cca-miR399 | MIMAT0024533 | |
| Glycine max | gma-miR399j | MIMAT0041664 | |
| Glycine max | gma-miR399k | MIMAT0041672 | |
| Gossypium hirsutum | ghr-miR399d | MIMAT0014350 | |
| View More Rows | |||
| Medicago truncatula | mtr-miR399c | MIMAT0001650 | |
| Medicago truncatula | mtr-miR399h | MIMAT0011075 | |
| Medicago truncatula | mtr-miR399i | MIMAT0011076 | |
Stem-loop Details
Arabidopsis
Arabidopsis lyrata
Cynara cardunculus
Glycine max
Gossypium hirsutum
Medicago truncatula

Genomic Map