Assay Details
Gene Family ID: | MIPF0000015, MIR399 |
Mature miRNA Sequence: | UGCCAAAGGAGAUUUGCCCUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Arabidopsis | ath-miR399a | MIMAT0000951 |
Arabidopsis lyrata | aly-miR399f-3p | MIMAT0017582 |
Cynara cardunculus | cca-miR399 | MIMAT0024533 |
Glycine max | gma-miR399j | MIMAT0041664 |
Glycine max | gma-miR399k | MIMAT0041672 |
Gossypium hirsutum | ghr-miR399d | MIMAT0014350 |
View More Rows | ||
Medicago truncatula | mtr-miR399c | MIMAT0001650 |
Medicago truncatula | mtr-miR399h | MIMAT0011075 |
Medicago truncatula | mtr-miR399i | MIMAT0011076 |
Stem-loop Details
Arabidopsis
Arabidopsis lyrata
Cynara cardunculus
Glycine max
Gossypium hirsutum
Medicago truncatula
