Assay Details
Mature miRNA Sequence: | UAAAGCUAGAUAACCGAAAGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Alligator mississippiensis | ami-miR-9-3p | MIMAT0038028 |
| Gadus morhua | gmo-miR-9-3p | MIMAT0044077 |
| Xenopus tropicalis | xtr-miR-9-3p | MIMAT0003650 |
| Xenopus tropicalis | xtr-miR-9a-3p | MIMAT0003554 |
| Zebrafish | dre-miR-9-3p | MIMAT0003156 |
Stem-loop Details
Alligator mississippiensis
Gadus morhua
Xenopus tropicalis
Zebrafish
More Information
Related Products
miRBase Alias:
Human: hsa-miR-9* (v9.2)
Mouse: mmu-miR-9* (v9.2)
Danio rerio: dre-miR-9* (v19)
Monodelphis domestica: mdo-miR-9* (v19)
Monodelphis domestica: mdo-miR-9-3p (v19)
Xenopus tropicalis: xtr-miR-9a* (v19)
Xenopus tropicalis: xtr-miR-9* (v18)
Mouse: mmu-miR-9* (v9.2)
Danio rerio: dre-miR-9* (v19)
Monodelphis domestica: mdo-miR-9* (v19)
Monodelphis domestica: mdo-miR-9-3p (v19)
Xenopus tropicalis: xtr-miR-9a* (v19)
Xenopus tropicalis: xtr-miR-9* (v18)

Genomic Map