Assay Details
Mature miRNA Sequence: | UAAAGCUAGAUAACCGAAAGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Alligator mississippiensis | ami-miR-9-3p | MIMAT0038028 |
| Gadus morhua | gmo-miR-9-3p | MIMAT0044077 |
| Xenopus tropicalis | xtr-miR-9-3p | MIMAT0003650 |
| Xenopus tropicalis | xtr-miR-9a-3p | MIMAT0003554 |
| Zebrafish | dre-miR-9-3p | MIMAT0003156 |
Stem-loop Details
Alligator mississippiensis
| Stem-loop ID | ami-mir-9-1 |
| Stem-loop Accession # | MI0029659 |
| Stem-loop Sequence | GGGUUGGUUGUUAUCUUUGGUUAUCUAGCUGUAUGAGUGGUGUCAAUUCUUCAUAAAGCUAGAUAACCGAAAGUAAAAAUAACCCCAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038028
miRBase ID: ami-miR-9-3p Mature miRNA Sequence: UAAAGCUAGAUAACCGAAAGU Chromosome Location: NA |
| Stem-loop ID | ami-mir-9-2 |
| Stem-loop Accession # | MI0029660 |
| Stem-loop Sequence | GAAGCGAGUUGUUAUCUUUGGUUAUCUAGCUGUAUGAGUGUUUUGGUCUUCAUAAAGCUAGAUAACCGAAAGUAAAAACUCCUUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038028
miRBase ID: ami-miR-9-3p Mature miRNA Sequence: UAAAGCUAGAUAACCGAAAGU Chromosome Location: NA |
| Stem-loop ID | ami-mir-9-3 |
| Stem-loop Accession # | MI0029661 |
| Stem-loop Sequence | AGGGCUGUUUCUGUCUUUGGUUAUCUAGCUGUAUGAGUGUUCUCGAGCCGUCAUAAAGCUAGAUAACCGAAAGUAGAAAUGACUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038028
miRBase ID: ami-miR-9-3p Mature miRNA Sequence: UAAAGCUAGAUAACCGAAAGU Chromosome Location: NA |
| Stem-loop ID | gmo-mir-9-1 |
| Stem-loop Accession # | MI0036020 |
| Stem-loop Sequence | UCUUUGGUUAUCUAGCUGUAUGAGUGGCAUACAUUCUUCAUAAAGCUAGAUAACCGAAAGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044077
miRBase ID: gmo-miR-9-3p Mature miRNA Sequence: UAAAGCUAGAUAACCGAAAGU Chromosome Location: NA |
| Stem-loop ID | gmo-mir-9-2 |
| Stem-loop Accession # | MI0036170 |
| Stem-loop Sequence | UCUUUGGUUAUCUAGCUGUAUGAGUGGUGUACAUUCUUCAUAAAGCUAGAUAACCGAAAGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044077
miRBase ID: gmo-miR-9-3p Mature miRNA Sequence: UAAAGCUAGAUAACCGAAAGU Chromosome Location: NA |
| Stem-loop ID | gmo-mir-9-7 |
| Stem-loop Accession # | MI0036101 |
| Stem-loop Sequence | UCUUUGGUUAUCUAGCUGUAUGAGUUUUAAAUCCCUGUCAUAAAGCUAGAUAACCGAAAGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044077
miRBase ID: gmo-miR-9-3p Mature miRNA Sequence: UAAAGCUAGAUAACCGAAAGU Chromosome Location: NA |
| Stem-loop ID | xtr-mir-9-3 |
| Stem-loop Accession # | MI0004891 |
| Stem-loop Sequence | GUUUCUAUCUUUGGUUAUCUAGCUGUAUGAGUGUAAAUAAGCCGUCAUAAAGCUAGAUAACCGAAAGUAGGAAUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003650
miRBase ID: xtr-miR-9-3p Mature miRNA Sequence: UAAAGCUAGAUAACCGAAAGU Chromosome Location: NA |
| Stem-loop ID | xtr-mir-9a-1 |
| Stem-loop Accession # | MI0004793 |
| Stem-loop Sequence | GGGGUUGGUUGUUAUCUUUGGUUAUCUAGCUGUAUGAGUGUUGUCAAUCCUUCAUAAAGCUAGAUAACCGAAAGUAAAAAUAACCCCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003554
miRBase ID: xtr-miR-9a-3p Mature miRNA Sequence: UAAAGCUAGAUAACCGAAAGU Chromosome Location: NA |
| Stem-loop ID | xtr-mir-9a-2 |
| Stem-loop Accession # | MI0004794 |
| Stem-loop Sequence | GGAAGUGGUUGUUAUCUUUGGUUAUCUAGCUGUAUGAGUGUAUUGGUUUUCAUAAAGCUAGAUAACCGAAAGUAAAAACUCCUUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003554
miRBase ID: xtr-miR-9a-3p Mature miRNA Sequence: UAAAGCUAGAUAACCGAAAGU Chromosome Location: NA |
| Stem-loop ID | dre-mir-9-1 |
| Stem-loop Accession # | MI0001880 |
| Stem-loop Sequence | GGGGUUGGCUGUUAUCUUUGGUUAUCUAGCUGUAUGAGUGUUAUUCAUUCUUCAUAAAGCUAGAUAACCGAAAGUAACAAGAAUCCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003156
miRBase ID: dre-miR-9-3p Mature miRNA Sequence: UAAAGCUAGAUAACCGAAAGU Chromosome Location: NA |
| Stem-loop ID | dre-mir-9-2 |
| Stem-loop Accession # | MI0001881 |
| Stem-loop Sequence | GCUCUCCUCUGGUGACUUUUGCACUUGGAGGCGUGUUGUUAUCUUUGGUUAUCUAGCUGUAUGAGUGUGCUGGCCGUCAUAAAGCUAGAUAACCGAAAGUAAGAGCCGCUUCUAUCAGCAACGUAUGGAGCAUUGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003156
miRBase ID: dre-miR-9-3p Mature miRNA Sequence: UAAAGCUAGAUAACCGAAAGU Chromosome Location: NA |
| Stem-loop ID | dre-mir-9-3 |
| Stem-loop Accession # | MI0001883 |
| Stem-loop Sequence | UCAGGGGGUUGGUUUCUCUCUUUGGUUAUCUAGCUGUAUGAGUUAUAACACUGUCAUAAAGCUAGAUAACCGAAAGUAGAAAUAAUUCCCAAAAUCUUUGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003156
miRBase ID: dre-miR-9-3p Mature miRNA Sequence: UAAAGCUAGAUAACCGAAAGU Chromosome Location: NA |
| Stem-loop ID | dre-mir-9-5 |
| Stem-loop Accession # | MI0001884 |
| Stem-loop Sequence | GGAAGCGAGUUGUUAUCUUUGGUUAUCUAGCUGUAUGAGUAUUUUGCACUUCAUAAAGCUAGAUAACCGAAAGUAAAAACUGCCUCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003156
miRBase ID: dre-miR-9-3p Mature miRNA Sequence: UAAAGCUAGAUAACCGAAAGU Chromosome Location: NA |
| Stem-loop ID | dre-mir-9-6 |
| Stem-loop Accession # | MI0001885 |
| Stem-loop Sequence | GGAGGUAGUUGCUAUCUUUGGUUAUCUAGCUGUAUGAGUGUUUAUCUGCCUUCAUAAAGCUAGAUAACCGAAAGUAGAAAUGUCCUCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003156
miRBase ID: dre-miR-9-3p Mature miRNA Sequence: UAAAGCUAGAUAACCGAAAGU Chromosome Location: NA |
More Information
Related Products
miRBase Alias:
Danio rerio: dre-miR-9* (v19)
Monodelphis domestica: mdo-miR-9* (v19)
Monodelphis domestica: mdo-miR-9-3p (v19)
Xenopus tropicalis: xtr-miR-9a* (v19)
Xenopus tropicalis: xtr-miR-9* (v18)
Monodelphis domestica: mdo-miR-9* (v19)
Monodelphis domestica: mdo-miR-9-3p (v19)
Xenopus tropicalis: xtr-miR-9a* (v19)
Xenopus tropicalis: xtr-miR-9* (v18)

Genomic Map