Assay Details
Gene Family ID: | MIPF0000149, mir-448 |
Mature miRNA Sequence: | UUGCAUAUGUAGGAUGUCCCAU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-448 | MIMAT0001532 | |
| Mouse | mmu-miR-448-3p | MIMAT0001533 | |
| Bovine | bta-miR-448 | MIMAT0009319 | |
| Dog | cfa-miR-448 | MIMAT0001535 | |
| Horse | eca-miR-448 | MIMAT0013218 | |
| Pan troglodytes | ptr-miR-448 | MIMAT0008148 | |
| View More Rows | |||
| Pongo pygmaeus | ppy-miR-448 | MIMAT0015890 | |
| Rhesus monkey | mml-miR-448 | MIMAT0006329 | |
Stem-loop Details
Human
Mouse
Bovine
Dog
Horse
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Mouse: mmu-miR-448 (v15)
Rat: rno-miR-448 (v15)
Rat: rno-miR-448 (v15)

Genomic Map