Assay Details
Gene Family ID: | MIPF0000149, mir-448 |
Mature miRNA Sequence: | UUGCAUAUGUAGGAUGUCCCAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-448 | MIMAT0001532 |
| Mouse | mmu-miR-448-3p | MIMAT0001533 |
| Bovine | bta-miR-448 | MIMAT0009319 |
| Dog | cfa-miR-448 | MIMAT0001535 |
| Horse | eca-miR-448 | MIMAT0013218 |
| Pan troglodytes | ptr-miR-448 | MIMAT0008148 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-448 | MIMAT0015890 |
| Rhesus monkey | mml-miR-448 | MIMAT0006329 |
Stem-loop Details
Human
Mouse
Bovine
Dog
Horse
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-448 (v15)
Rat: rno-miR-448 (v15)
Rat: rno-miR-448 (v15)

Genomic Map