Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | UCUCUGGAGGGAAGCACUUUCUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-518c-5p | MIMAT0002847 |
| Gorilla gorilla | ggo-miR-518c-5p | MIMAT0036421 |
Stem-loop Details
Human
Gorilla gorilla

Genomic Map