Assay Details
Amplicon Length: | 88 |
Chromosome Location: | Chr. 19 - 53708437 - 53708437 [-] on 53708437 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-518c-5p | MIMAT0002847 |
| Human | hsa-miR-518c-3p | MIMAT0002848 |
Stem-loop Details
Human

Genomic Map