Assay Details
Mature miRNA Sequence: | UCGGAUCCGUCUGAGCUUGGC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Callithrix jacchus | cja-miR-127 | MIMAT0039469 |
Gorilla gorilla | ggo-miR-127 | MIMAT0024094 |
Stem-loop Details
Callithrix jacchus
Gorilla gorilla
