Assay Details
Mature miRNA Sequence: | UCGGAUCCGUCUGAGCUUGGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Callithrix jacchus | cja-miR-127 | MIMAT0039469 |
| Gorilla gorilla | ggo-miR-127 | MIMAT0024094 |
Stem-loop Details
Callithrix jacchus
Gorilla gorilla

Genomic Map