Assay Details
Gene Family ID: | MIPF0000085, mir-140 |
Mature miRNA Sequence: | CAGUGGUUUUACCCUAUGGUAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-140-5p | MIMAT0000431 | |
| Mouse | mmu-miR-140-5p | MIMAT0000151 | |
| Rat | rno-miR-140-5p | MIMAT0000573 | |
| Alligator mississippiensis | ami-miR-140-5p | MIMAT0038160 | |
| Chrysemys picta | cpi-miR-140-5p | MIMAT0037764 | |
| Columba livia | cli-miR-140-5p | MIMAT0038527 | |
| View More Rows | |||
| Cricetulus griseus | cgr-miR-140-5p | MIMAT0023767 | |
| Cyprinus carpio | ccr-miR-140-5p | MIMAT0026220 | |
| Dasypus novemcinctus | dno-miR-140-5p | MIMAT0047653 | |
| Eptesicus fuscus | efu-miR-140 | MIMAT0035193 | |
| Fugu rubripes | fru-miR-140 | MIMAT0003024 | |
| Goat | chi-miR-140-5p | MIMAT0035960 | |
| Guinea pig | cpo-miR-140-5p | MIMAT0047004 | |
| Horse | eca-miR-140-5p | MIMAT0012926 | |
| Monodelphis domestica | mdo-miR-140-5p | MIMAT0012745 | |
| Ophiophagus hannah | oha-miR-140-5p | MIMAT0036719 | |
| Oryzias latipes | ola-miR-140-5p | MIMAT0022552 | |
| Paralichthys olivaceus | pol-miR-140-5p | MIMAT0025434 | |
| Petromyzon marinus | pma-miR-140 | MIMAT0019471 | |
| Pongo pygmaeus | ppy-miR-140-5p | MIMAT0015763 | |
| Pteropus alecto | pal-miR-140-5p | MIMAT0039778 | |
| Rabbit | ocu-miR-140-5p | MIMAT0048241 | |
| Rhesus monkey | mml-miR-140-5p | MIMAT0006197 | |
| Salmo salar | ssa-miR-140-5p | MIMAT0032359 | |
| Taeniopygia guttata | tgu-miR-140-5p | MIMAT0014557 | |
| Tetraodon nigroviridis | tni-miR-140 | MIMAT0003025 | |
| Zebrafish | dre-miR-140-5p | MIMAT0001836 | |
Stem-loop Details
Human
Mouse
Rat
Alligator mississippiensis
Chrysemys picta
Columba livia
Cricetulus griseus
Cyprinus carpio
Dasypus novemcinctus
Eptesicus fuscus
Fugu rubripes
Goat
Guinea pig
Horse
Monodelphis domestica
Ophiophagus hannah
Oryzias latipes
Paralichthys olivaceus
Petromyzon marinus
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
Salmo salar
Taeniopygia guttata
Tetraodon nigroviridis
Zebrafish
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Mouse: mmu-miR-140 (v17)
Rat: rno-miR-140 (v18)
Danio rerio: dre-miR-140 (v18)
Monodelphis domestica: mdo-miR-140 (v19)
Trabulsiella guamensis ATCC 49490: tgu-miR-140 (v18)
Rat: rno-miR-140 (v18)
Danio rerio: dre-miR-140 (v18)
Monodelphis domestica: mdo-miR-140 (v19)
Trabulsiella guamensis ATCC 49490: tgu-miR-140 (v18)

Genomic Map