Assay Details
Amplicon Length: | 61 |
Chromosome Location: | Chr. 19 - 39609201 - 39609201 [-] on 39609201 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-140-5p | MIMAT0000573 |
| Rat | rno-miR-140-3p | MIMAT0000574 |
Stem-loop Details
Rat

Genomic Map