Assay Details
Amplicon Length: | 99 |
Chromosome Location: | Chr. 8 - 107550843 - 107550843 [-] on 107550843 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-140-5p | MIMAT0000151 |
| Mouse | mmu-miR-140-3p | MIMAT0000152 |
Stem-loop Details
Mouse

Genomic Map