Assay Details
Amplicon Length: | 86 |
Chromosome Location: | Chr. 16 - 69932883 - 69932883 [-] on 69932883 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-140-3p | MIMAT0004597 |
| Human | hsa-miR-140-5p | MIMAT0000431 |
Stem-loop Details
Human

Genomic Map