Assay Details
Gene Family ID: | MIPF0000085, mir-140 |
Mature miRNA Sequence: | CAGUGGUUUUACCCUAUGGUAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-140-5p | MIMAT0000431 |
| Mouse | mmu-miR-140-5p | MIMAT0000151 |
| Rat | rno-miR-140-5p | MIMAT0000573 |
| Alligator mississippiensis | ami-miR-140-5p | MIMAT0038160 |
| Chrysemys picta | cpi-miR-140-5p | MIMAT0037764 |
| Columba livia | cli-miR-140-5p | MIMAT0038527 |
| View More Rows | ||
| Cricetulus griseus | cgr-miR-140-5p | MIMAT0023767 |
| Cyprinus carpio | ccr-miR-140-5p | MIMAT0026220 |
| Dasypus novemcinctus | dno-miR-140-5p | MIMAT0047653 |
| Eptesicus fuscus | efu-miR-140 | MIMAT0035193 |
| Fugu rubripes | fru-miR-140 | MIMAT0003024 |
| Goat | chi-miR-140-5p | MIMAT0035960 |
| Guinea pig | cpo-miR-140-5p | MIMAT0047004 |
| Horse | eca-miR-140-5p | MIMAT0012926 |
| Monodelphis domestica | mdo-miR-140-5p | MIMAT0012745 |
| Ophiophagus hannah | oha-miR-140-5p | MIMAT0036719 |
| Oryzias latipes | ola-miR-140-5p | MIMAT0022552 |
| Paralichthys olivaceus | pol-miR-140-5p | MIMAT0025434 |
| Petromyzon marinus | pma-miR-140 | MIMAT0019471 |
| Pongo pygmaeus | ppy-miR-140-5p | MIMAT0015763 |
| Pteropus alecto | pal-miR-140-5p | MIMAT0039778 |
| Rabbit | ocu-miR-140-5p | MIMAT0048241 |
| Rhesus monkey | mml-miR-140-5p | MIMAT0006197 |
| Salmo salar | ssa-miR-140-5p | MIMAT0032359 |
| Taeniopygia guttata | tgu-miR-140-5p | MIMAT0014557 |
| Tetraodon nigroviridis | tni-miR-140 | MIMAT0003025 |
| Zebrafish | dre-miR-140-5p | MIMAT0001836 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-140 |
| Stem-loop Accession # | MI0000456 |
| Stem-loop Sequence | UGUGUCUCUCUCUGUGUCCUGCCAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGACAGGAUACCGGGGCACC |
| Chromosome Location | Chr. 16 - 69933081 - 69933180 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000431
miRBase ID: hsa-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: Chr. 16 - 69933081 - 69933180 [+] on Build GRCh38 |
| Stem-loop ID | mmu-mir-140 |
| Stem-loop Accession # | MI0000165 |
| Stem-loop Sequence | CCUGCCAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGACAGG |
| Chromosome Location | Chr. 8 - 107551244 - 107551313 [+] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000151
miRBase ID: mmu-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: Chr. 8 - 107551244 - 107551313 [+] on Build GRCm38 |
| Stem-loop ID | rno-mir-140 |
| Stem-loop Accession # | MI0000611 |
| Stem-loop Sequence | GUGUCUCUCUCUGUGUCCUGCCAGUGGUUUUACCCUAUGGUAGGUUACAUCAUGCUGUUCUACCACAGGGUAGAACCACGGACAGGAUACUGGAGCACC |
| Chromosome Location | Chr. 19 - 39608951 - 39609049 [+] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000573
miRBase ID: rno-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: Chr. 19 - 39608951 - 39609049 [+] on Build Rnor_6.0 |
| Stem-loop ID | ami-mir-140 |
| Stem-loop Accession # | MI0029745 |
| Stem-loop Sequence | GCUCUCUCUCUGUGUCCUGCCAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGACGGGAUACCGGGGCGCG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038160
miRBase ID: ami-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | cpi-mir-140 |
| Stem-loop Accession # | MI0029483 |
| Stem-loop Sequence | UCUCUGUGUCCUGCCAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGACAGGAUACCGGGCGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0037764
miRBase ID: cpi-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | cli-mir-140 |
| Stem-loop Accession # | MI0029976 |
| Stem-loop Sequence | UCCGUGUCCUGCCAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGACGGGAUGCUGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038527
miRBase ID: cli-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | cgr-mir-140 |
| Stem-loop Accession # | MI0020402 |
| Stem-loop Sequence | GUGUCCUGCCAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGACAGGAUACCGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0023767
miRBase ID: cgr-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | ccr-mir-140 |
| Stem-loop Accession # | MI0023326 |
| Stem-loop Sequence | GUCUCCUGUGUCCUGUCAGUGGUUUUACCCUAUGGUAGGUUACGUCAUACUGUUCUACCACAGGGUAGAACCACGGACCGGAUGUCUGGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0026220
miRBase ID: ccr-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | dno-mir-140 |
| Stem-loop Accession # | MI0038989 |
| Stem-loop Sequence | CAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGAC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047653
miRBase ID: dno-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | efu-mir-140 |
| Stem-loop Accession # | MI0028907 |
| Stem-loop Sequence | UGUGUCCUGCCAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGACAGGAUACCG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0035193
miRBase ID: efu-miR-140 Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | fru-mir-140 |
| Stem-loop Accession # | MI0003352 |
| Stem-loop Sequence | GUCAGUGGUUUUACCCUAUGGUAGGUGACAUCAUGCUGUUCUACCACAGGGUAGAACCACGGACGGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003024
miRBase ID: fru-miR-140 Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | chi-mir-140 |
| Stem-loop Accession # | MI0030627 |
| Stem-loop Sequence | UGUGUGUCUGUCUCUCUGUGUCCUGCCAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGACAGGAUACCGGGGCACCCUCUGCAUCG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0035960
miRBase ID: chi-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | cpo-mir-140 |
| Stem-loop Accession # | MI0038647 |
| Stem-loop Sequence | CAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGAC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047004
miRBase ID: cpo-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | eca-mir-140 |
| Stem-loop Accession # | MI0012682 |
| Stem-loop Sequence | CCAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0012926
miRBase ID: eca-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | mdo-mir-140 |
| Stem-loop Accession # | MI0012501 |
| Stem-loop Sequence | CCUGCCAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGACAGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0012745
miRBase ID: mdo-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | oha-mir-140 |
| Stem-loop Accession # | MI0031354 |
| Stem-loop Sequence | GGUGUCCCGCCAGUGGUUUUACCCUAUGGUAGGUGACCUGGUGCUGUUCUACCACAGGGUAGAACCACGGACGGGAUGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036719
miRBase ID: oha-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | ola-mir-140 |
| Stem-loop Accession # | MI0019434 |
| Stem-loop Sequence | UGUCUCCGGUGUCACGUCAGUGGUUUUACCCUAUGGUAGGUGACAUCAUGCUGUUCUACCACAGGGUAGAACCACGGACGGGAUGUUGGGAGGUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0022552
miRBase ID: ola-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | pol-mir-140 |
| Stem-loop Accession # | MI0022200 |
| Stem-loop Sequence | CAGUGGUUUUACCCUAUGGUAGGUGACAUCAUGCUGUUCUACCACAGGGUAGAACCACGGAC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0025434
miRBase ID: pol-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | pma-mir-140 |
| Stem-loop Accession # | MI0017077 |
| Stem-loop Sequence | UCGACCGCUCAGUGGUUUUACCCUAUGGUAGCUUUGAAUGGCAGACUCUACCACAUGGUAGAAUCACGGCCCGGUGAC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0019471
miRBase ID: pma-miR-140 Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | ppy-mir-140 |
| Stem-loop Accession # | MI0014832 |
| Stem-loop Sequence | UGUGUCUCUCUCUGUGUCCUGCCAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGACAGGAUACCGGGGCACC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0015763
miRBase ID: ppy-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | pal-mir-140 |
| Stem-loop Accession # | MI0032323 |
| Stem-loop Sequence | CAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039778
miRBase ID: pal-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | ocu-mir-140 |
| Stem-loop Accession # | MI0039298 |
| Stem-loop Sequence | CAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGAC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048241
miRBase ID: ocu-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | mml-mir-140 |
| Stem-loop Accession # | MI0007630 |
| Stem-loop Sequence | UGUGUCUCUCUCUGUGUCCUGCCAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGACAGGAUACCGGGGCACC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006197
miRBase ID: mml-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | ssa-mir-140 |
| Stem-loop Accession # | MI0026499 |
| Stem-loop Sequence | CAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032359
miRBase ID: ssa-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | tgu-mir-140 |
| Stem-loop Accession # | MI0013774 |
| Stem-loop Sequence | CGUGUCCUGCCAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGAC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0014557
miRBase ID: tgu-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | tni-mir-140 |
| Stem-loop Accession # | MI0003353 |
| Stem-loop Sequence | GUCAGUGGUUUUACCCUAUGGUAGGUGACAUCAUGCUGUUCUACCACAGGGUAGAACCACGGACGGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003025
miRBase ID: tni-miR-140 Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
| Stem-loop ID | dre-mir-140 |
| Stem-loop Accession # | MI0002003 |
| Stem-loop Sequence | GUGUUUGUCUCCUGUGUCCCGUCAGUGGUUUUACCCUAUGGUAGGUUACGUCAUGCUGUUCUACCACAGGGUAGAACCACGGACGGGAUGUCUGGAGGUGUCUGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001836
miRBase ID: dre-miR-140-5p Mature miRNA Sequence: CAGUGGUUUUACCCUAUGGUAG Chromosome Location: NA |
More Information
Related Products

Genomic Map