Assay Details
Gene Family ID: | MIPF0000201, mir-485 |
Mature miRNA Sequence: | GUCAUACACGGCUCUCCUCUCU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-485-3p | MIMAT0002176 | |
| Dasypus novemcinctus | dno-miR-485-3p | MIMAT0047911 | |
| Eptesicus fuscus | efu-miR-485 | MIMAT0035038 | |
| Horse | eca-miR-485-3p | MIMAT0013160 | |
| Pan troglodytes | ptr-miR-485 | MIMAT0008159 | |
| Pongo pygmaeus | ppy-miR-485-3p | MIMAT0015903 | |
| View More Rows | |||
| Pteropus alecto | pal-miR-485-3p | MIMAT0039879 | |
| Rabbit | ocu-miR-485-3p | MIMAT0048468 | |
| Rhesus monkey | mml-miR-485-3p | MIMAT0006343 | |
| Sheep | oar-miR-485-3p | MIMAT0019313 | |
Stem-loop Details
Human
Dasypus novemcinctus
Eptesicus fuscus
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
Sheep

Genomic Map