Assay Details
Amplicon Length: | 69 |
Chromosome Location: | Chr. 14 - 101055658 - 101055658 [+] on 101055658 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-485-5p | MIMAT0002175 |
| Human | hsa-miR-485-3p | MIMAT0002176 |
Stem-loop Details
Human

Genomic Map