Assay Details
Gene Family ID: | MIPF0000557, mir-619 |
Mature miRNA Sequence: | GACCUGGACAUGUUUGUGCCCAGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-619-3p | MIMAT0003288 |
| Pan troglodytes | ptr-miR-619 | MIMAT0008281 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map