Assay Details
Amplicon Length: | 72 |
Chromosome Location: | Chr. 12 - 108836702 - 108836702 [-] on 108836702 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-619-5p | MIMAT0026622 |
| Human | hsa-miR-619-3p | MIMAT0003288 |
Stem-loop Details
Human

Genomic Map