Assay Details
Gene Family ID: | MIPF0000605, mir-646 | 
Mature miRNA Sequence: | AAGCAGCUGCCUCUGAGGC | 
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-646 | MIMAT0003316 | 
| Pan troglodytes | ptr-miR-646 | MIMAT0008299 | 
Stem-loop Details
Human
Pan troglodytes

Genomic Map