Assay Details
Gene Family ID: | MIPF0000605, mir-646 |
Mature miRNA Sequence: | AAGCAGCUGCCUCUGAGGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-646 | MIMAT0003316 |
Pan troglodytes | ptr-miR-646 | MIMAT0008299 |
Stem-loop Details
Human
Pan troglodytes
