Assay Details
Gene Family ID: | MIPF0000033, mir-10 |
Mature miRNA Sequence: | UACCCUGUAGAACCGAAUUUGUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-10b-5p | MIMAT0000254 | |
| Mouse | mmu-miR-10b-5p | MIMAT0000208 | |
| Bovine | bta-miR-10b | MIMAT0003839 | |
| Fugu rubripes | fru-miR-10b | MIMAT0002963 | |
| Horse | eca-miR-10b | MIMAT0013090 | |
| Pan troglodytes | ptr-miR-10b | MIMAT0007945 | |
| View More Rows | |||
| Pongo pygmaeus | ppy-miR-10b | MIMAT0015730 | |
| Rhesus monkey | mml-miR-10b-5p | MIMAT0006162 | |
| Tetraodon nigroviridis | tni-miR-10b | MIMAT0002964 | |
| Tupaia chinensis | tch-miR-10b-5p | MIMAT0036471 | |
| Zebrafish | dre-miR-10b-5p | MIMAT0001268 | |
Stem-loop Details
Human
Mouse
Bovine
Fugu rubripes
Horse
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
Tetraodon nigroviridis
Tupaia chinensis
Zebrafish
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products
miRBase Alias:
Human: hsa-miR-10b (v17)
Mouse: mmu-miR-10b (v17)
Danio rerio: dre-miR-10b (v20)
Macaca mulatta: mml-miR-10b (v19)
Mouse: mmu-miR-10b (v17)
Danio rerio: dre-miR-10b (v20)
Macaca mulatta: mml-miR-10b (v19)

Genomic Map