Assay Details
Amplicon Length: | 62 |
Chromosome Location: | Chr. 2 - 74726573 - 74726573 [+] on 74726573 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-10b-5p | MIMAT0000208 |
| Mouse | mmu-miR-10b-3p | MIMAT0004538 |
Stem-loop Details
Mouse

Genomic Map