Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | CACAUUACACGGUCGACCUCU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-323a-3p | MIMAT0000755 | |
| Mouse | mmu-miR-323-3p | MIMAT0000551 | |
| Rat | rno-miR-323-3p | MIMAT0000550 | |
| Dasypus novemcinctus | dno-miR-323a-3p | MIMAT0047800 | |
| Dog | cfa-miR-323 | MIMAT0006718 | |
| Goat | chi-miR-323a-3p | MIMAT0036133 | |
| View More Rows | |||
| Guinea pig | cpo-miR-323a-3p | MIMAT0047155 | |
| Horse | eca-miR-323-3p | MIMAT0013132 | |
| Pan paniscus | ppa-miR-323 | MIMAT0049044 | |
| Pan troglodytes | ptr-miR-323 | MIMAT0008097 | |
| Pongo pygmaeus | ppy-miR-323-3p | MIMAT0015826 | |
| Pteropus alecto | pal-miR-323-3p | MIMAT0039922 | |
| Rhesus monkey | mml-miR-323a-3p | MIMAT0006265 | |
| Sheep | oar-miR-323a-3p | MIMAT0019260 | |
Stem-loop Details
Human
Mouse
Rat
Dasypus novemcinctus
Dog
Goat
Guinea pig
Horse
Pan paniscus
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rhesus monkey
Sheep
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Human: hsa-miR-323-3p (v17)
Rat: rno-miR-323 (v18)
Macaca mulatta: mml-miR-323-3p (v19)
Rat: rno-miR-323 (v18)
Macaca mulatta: mml-miR-323-3p (v19)

Genomic Map