Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | CACAUUACACGGUCGACCUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-323a-3p | MIMAT0000755 |
| Mouse | mmu-miR-323-3p | MIMAT0000551 |
| Rat | rno-miR-323-3p | MIMAT0000550 |
| Dasypus novemcinctus | dno-miR-323a-3p | MIMAT0047800 |
| Dog | cfa-miR-323 | MIMAT0006718 |
| Goat | chi-miR-323a-3p | MIMAT0036133 |
| View More Rows | ||
| Guinea pig | cpo-miR-323a-3p | MIMAT0047155 |
| Horse | eca-miR-323-3p | MIMAT0013132 |
| Pan paniscus | ppa-miR-323 | MIMAT0049044 |
| Pan troglodytes | ptr-miR-323 | MIMAT0008097 |
| Pongo pygmaeus | ppy-miR-323-3p | MIMAT0015826 |
| Pteropus alecto | pal-miR-323-3p | MIMAT0039922 |
| Rhesus monkey | mml-miR-323a-3p | MIMAT0006265 |
| Sheep | oar-miR-323a-3p | MIMAT0019260 |
Stem-loop Details
Human
Mouse
Rat
Dasypus novemcinctus
Dog
Goat
Guinea pig
Horse
Pan paniscus
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rhesus monkey
Sheep
More Information
Related Products
miRBase Alias:
Human: hsa-miR-323-3p (v17)
Rat: rno-miR-323 (v18)
Macaca mulatta: mml-miR-323-3p (v19)
Rat: rno-miR-323 (v18)
Macaca mulatta: mml-miR-323-3p (v19)

Genomic Map