Assay Details
Gene Family ID: | MIPF0000085, mir-140 |
Mature miRNA Sequence: | UACCACAGGGUAGAACCACGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-140-3p | MIMAT0004597 | |
| Mouse | mmu-miR-140-3p | MIMAT0000152 | |
| Rat | rno-miR-140-3p | MIMAT0000574 | |
| Anolis carolinensis | aca-miR-140-3p | MIMAT0021766 | |
| Horse | eca-miR-140-3p | MIMAT0012927 | |
| Pongo pygmaeus | ppy-miR-140-3p | MIMAT0015764 | |
| View More Rows | |||
| Rhesus monkey | mml-miR-140-3p | MIMAT0006198 | |
Stem-loop Details
Human
Mouse
Rat
Anolis carolinensis
Horse
Pongo pygmaeus
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool B v3.0Megaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Mouse: mmu-miR-140* (v17)
Rat: rno-miR-140* (v18)
Rat: rno-miR-140* (v18)

Genomic Map