Assay Details
Gene Family ID: | MIPF0000085, mir-140 |
Mature miRNA Sequence: | UACCACAGGGUAGAACCACGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-140-3p | MIMAT0004597 |
| Mouse | mmu-miR-140-3p | MIMAT0000152 |
| Rat | rno-miR-140-3p | MIMAT0000574 |
| Anolis carolinensis | aca-miR-140-3p | MIMAT0021766 |
| Horse | eca-miR-140-3p | MIMAT0012927 |
| Pongo pygmaeus | ppy-miR-140-3p | MIMAT0015764 |
| View More Rows | ||
| Rhesus monkey | mml-miR-140-3p | MIMAT0006198 |
Stem-loop Details
Human
Mouse
Rat
Anolis carolinensis
Horse
Pongo pygmaeus
Rhesus monkey
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-140* (v17)
Rat: rno-miR-140* (v18)
Rat: rno-miR-140* (v18)

Genomic Map