Assay Details
Gene Family ID: | MIPF0000076, mir-190 |
Mature miRNA Sequence: | UGAUAUGUUUGAUAUUGGGUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Bovine | bta-miR-190b | MIMAT0009252 | |
| Dog | cfa-miR-190b | MIMAT0009873 | |
| Horse | eca-miR-190b | MIMAT0012958 | |
| Monodelphis domestica | mdo-miR-190b | MIMAT0012756 | |
| Pan troglodytes | ptr-miR-190b | MIMAT0008055 | |
| Pongo pygmaeus | ppy-miR-190b | MIMAT0015784 | |
| View More Rows | |||
| Rhesus monkey | mml-miR-190b | MIMAT0006222 | |
| Tupaia chinensis | tch-miR-190b | MIMAT0036501 | |
Stem-loop Details
Bovine
Dog
Horse
Monodelphis domestica
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
Tupaia chinensis
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool B v3.0Megaplex™ RT Primers, Human Pool B v3.0
Associated TaqMan® Array MicroRNA Cards:
TaqMan® Human MicroRNA B Cards v3.0TaqMan® Rodent MicroRNA B Cards v3.0
TaqMan® OpenArray® MicroRNA Panels:
TaqMan® OpenArray® Human MicroRNA PanelTaqMan® OpenArray® Rodent MicroRNA Panel
Related Products
miRBase Alias:
Human: hsa-miR-190b (v21)
Mouse: mmu-miR-190b-5p (v21)
Mouse: mmu-miR-190b (v17)
Mouse: mmu-miR-190b-5p (v21)
Mouse: mmu-miR-190b (v17)

Genomic Map