Assay Details
Gene Family ID: | MIPF0000076, mir-190 |
Mature miRNA Sequence: | UGAUAUGUUUGAUAUUGGGUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-190b | MIMAT0009252 |
| Dog | cfa-miR-190b | MIMAT0009873 |
| Horse | eca-miR-190b | MIMAT0012958 |
| Monodelphis domestica | mdo-miR-190b | MIMAT0012756 |
| Pan troglodytes | ptr-miR-190b | MIMAT0008055 |
| Pongo pygmaeus | ppy-miR-190b | MIMAT0015784 |
| View More Rows | ||
| Rhesus monkey | mml-miR-190b | MIMAT0006222 |
| Tupaia chinensis | tch-miR-190b | MIMAT0036501 |
Stem-loop Details
Bovine
Dog
Horse
Monodelphis domestica
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
Tupaia chinensis
More Information
Related Products
miRBase Alias:
Human: hsa-miR-190b (v21)
Mouse: mmu-miR-190b-5p (v21)
Mouse: mmu-miR-190b-5p (v21)

Genomic Map