Assay Details
Gene Family ID: | MIPF0000034, mir-130 |
Mature miRNA Sequence: | CAGUGCAAUGAUAUUGUCAAAGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-301b-3p | MIMAT0004958 | |
| Cricetulus griseus | cgr-miR-301b | MIMAT0041138 | |
| Dog | cfa-miR-301b | MIMAT0009854 | |
| Goat | chi-miR-301b | MIMAT0036120 | |
| Horse | eca-miR-301b-3p | MIMAT0013001 | |
| Pan troglodytes | ptr-miR-301b | MIMAT0008085 | |
| View More Rows | |||
| Pongo pygmaeus | ppy-miR-301b | MIMAT0015816 | |
| Rhesus monkey | mml-miR-301b | MIMAT0006258 | |
Stem-loop Details
Human
Cricetulus griseus
Dog
Goat
Horse
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

Genomic Map