Assay Details
Amplicon Length: | 56 |
Chromosome Location: | Chr. 22 - 21652779 - 21652779 [+] on 21652779 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-301b-3p | MIMAT0004958 |
| Human | hsa-miR-301b-5p | MIMAT0032026 |
Stem-loop Details
Human

Genomic Map