Assay Details
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | CACUAGAUUGUGAGCUCCUGGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-28-3p | MIMAT0004502 | |
| Rat | rno-miR-28-3p | MIMAT0004716 | |
| Cricetulus griseus | cgr-miR-28-3p | MIMAT0023879 | |
| Dasypus novemcinctus | dno-miR-28-3p | MIMAT0047548 | |
| Daubentonia madagascariensis | dma-miR-28 | MIMAT0049257 | |
| Dog | cfa-miR-28 | MIMAT0006675 | |
| View More Rows | |||
| Horse | eca-miR-28-3p | MIMAT0013093 | |
| Microcebus murinus | mmr-miR-28 | MIMAT0049146 | |
| Nomascus leucogenys | nle-miR-28 | MIMAT0049459 | |
| Pig | ssc-miR-28-3p | MIMAT0015211 | |
| Pteropus alecto | pal-miR-28-3p | MIMAT0039891 | |
| Rhesus monkey | mml-miR-28-3p | MIMAT0026575 | |
Stem-loop Details
Human
Rat
Cricetulus griseus
Dasypus novemcinctus
Daubentonia madagascariensis
Dog
Horse
Microcebus murinus
Nomascus leucogenys
Pig
Pteropus alecto
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool B v3.0
Related Products
miRBase Alias:
Rat: rno-miR-28* (v18)

Genomic Map