Assay Details
Amplicon Length: | 68 |
Chromosome Location: | Chr. 11 - 79380277 - 79380277 [+] on 79380277 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-28-5p | MIMAT0000800 |
| Rat | rno-miR-28-3p | MIMAT0004716 |
Stem-loop Details
Rat

Genomic Map