Assay Details
Gene Family ID: | MIPF0000417, mir-582 |
Mature miRNA Sequence: | UACAGUUGUUCAACCAGUUACU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-582-5p | MIMAT0012833 |
| Dog | cfa-miR-582 | MIMAT0009916 |
| Goat | chi-miR-582-5p | MIMAT0036279 |
| Pig | ssc-miR-582-5p | MIMAT0037466 |
Stem-loop Details
Rat
Dog
Goat
Pig
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-582-5p (v17)
Rat: rno-miR-582 (v18)
Rat: rno-miR-582 (v18)

Genomic Map