Assay Details
Gene Family ID: | MIPF0000417, mir-582 |
Mature miRNA Sequence: | UACAGUUGUUCAACCAGUUACU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-582-5p | MIMAT0012833 |
Dog | cfa-miR-582 | MIMAT0009916 |
Goat | chi-miR-582-5p | MIMAT0036279 |
Pig | ssc-miR-582-5p | MIMAT0037466 |
Stem-loop Details
Rat
Dog
Goat
Pig

More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-582-5p (v17)
Rat: rno-miR-582 (v18)
Rat: rno-miR-582 (v18)
