Assay Details
Gene Family ID: | MIPF0000621, mir-1251 |
Mature miRNA Sequence: | ACUCUAGCUGCCAAAGGCGCU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-1251-5p | MIMAT0005903 | |
Mouse | mmu-miR-1251-5p | MIMAT0014824 | |
Bovine | bta-miR-1251 | MIMAT0009963 | |
Callithrix jacchus | cja-miR-1251 | MIMAT0039364 | |
Dasypus novemcinctus | dno-miR-1251-5p | MIMAT0048033 | |
Guinea pig | cpo-miR-1251-5p | MIMAT0047407 | |
View More Rows | |||
Pan troglodytes | ptr-miR-1251 | MIMAT0007977 | |
Pongo pygmaeus | ppy-miR-1251 | MIMAT0016176 | |
Rabbit | ocu-miR-1251-5p | MIMAT0048587 | |
Rhesus monkey | mml-miR-1251-5p | MIMAT0028242 |
Stem-loop Details
Human
Mouse
Bovine
Callithrix jacchus
Dasypus novemcinctus
Guinea pig
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey

More Information
Related Products
miRBase Alias:
Human: hsa-miR-1251 (v19)
Mouse: mmu-miR-1251 (v17)
Mouse: mmu-miR-1251 (v17)
