Assay Details
Gene Family ID: | MIPF0000621, mir-1251 |
Mature miRNA Sequence: | ACUCUAGCUGCCAAAGGCGCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1251-5p | MIMAT0005903 |
Mouse | mmu-miR-1251-5p | MIMAT0014824 |
Bovine | bta-miR-1251 | MIMAT0009963 |
Callithrix jacchus | cja-miR-1251 | MIMAT0039364 |
Dasypus novemcinctus | dno-miR-1251-5p | MIMAT0048033 |
Guinea pig | cpo-miR-1251-5p | MIMAT0047407 |
View More Rows | ||
Pan troglodytes | ptr-miR-1251 | MIMAT0007977 |
Pongo pygmaeus | ppy-miR-1251 | MIMAT0016176 |
Rabbit | ocu-miR-1251-5p | MIMAT0048587 |
Rhesus monkey | mml-miR-1251-5p | MIMAT0028242 |
Stem-loop Details
Human
Mouse
Bovine
Callithrix jacchus
Dasypus novemcinctus
Guinea pig
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey
