Assay Details
Gene Family ID: | MIPF0000621, mir-1251 |
Mature miRNA Sequence: | ACUCUAGCUGCCAAAGGCGCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1251-5p | MIMAT0005903 |
| Mouse | mmu-miR-1251-5p | MIMAT0014824 |
| Bovine | bta-miR-1251 | MIMAT0009963 |
| Callithrix jacchus | cja-miR-1251 | MIMAT0039364 |
| Dasypus novemcinctus | dno-miR-1251-5p | MIMAT0048033 |
| Guinea pig | cpo-miR-1251-5p | MIMAT0047407 |
| View More Rows | ||
| Pan troglodytes | ptr-miR-1251 | MIMAT0007977 |
| Pongo pygmaeus | ppy-miR-1251 | MIMAT0016176 |
| Rabbit | ocu-miR-1251-5p | MIMAT0048587 |
| Rhesus monkey | mml-miR-1251-5p | MIMAT0028242 |
Stem-loop Details
Human
Mouse
Bovine
Callithrix jacchus
Dasypus novemcinctus
Guinea pig
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-1251 (v19)
Mouse: mmu-miR-1251 (v17)
Mouse: mmu-miR-1251 (v17)

Genomic Map