Assay Details
Gene Family ID: | MIPF0000673, mir-1182 |
Mature miRNA Sequence: | GAGGGUCUUGGGAGGGAUGUGAC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1182 | MIMAT0005827 |
| Pan troglodytes | ptr-miR-1182 | MIMAT0007950 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map