Assay Details
Amplicon Length: | 82 |
Chromosome Location: | Chr. 1 - 231020230 - 231020230 [-] on 231020230 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1182 | MIMAT0005827 |
Stem-loop Details
Human
